Product Details

SNP ID
rs4974602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1348603 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGTTCACTTCCTCCAAGGAAAA[C/T]GCCGGAGGGGATCTCTGCCGTCACC
Phenotype
MIM: 606801 MIM: 614632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAEA PubMed Links

Gene Details

Gene
MAEA
Gene Name
macrophage erythroblast attacher
There are no transcripts associated with this gene.

Gene
UVSSA
Gene Name
UV stimulated scaffold protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317934.1 Intron NP_001304863.1
NM_001317935.1 Intron NP_001304864.1
NM_020894.3 Intron NP_065945.2
XM_017008490.1 Intron XP_016863979.1
XM_017008491.1 Intron XP_016863980.1
XM_017008492.1 Intron XP_016863981.1
XM_017008493.1 Intron XP_016863982.1
XM_017008494.1 Intron XP_016863983.1
XM_017008495.1 Intron XP_016863984.1
XM_017008496.1 Intron XP_016863985.1
XM_017008497.1 Intron XP_016863986.1
XM_017008498.1 Intron XP_016863987.1
XM_017008499.1 Intron XP_016863988.1
XM_017008500.1 Intron XP_016863989.1

View Full Product Details