Product Details
- SNP ID
-
rs150380
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:36880769 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAGTTGTCATTTATAATAATTCTT[C/T]GTATTTTAAAACTAGTCCTTTTTCA
- Phenotype
-
MIM: 608667
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NIPBL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148337128] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NIPBL
- Gene Name
- NIPBL, cohesin loading factor
- Gene
- NIPBL-AS1
- Gene Name
- NIPBL antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details