Product Details

SNP ID
rs470171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102770721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGCAGGAAAAATTAACCATTTTG[C/G]CTCATAATACATTAGATGAATAATT
Phenotype
MIM: 185260
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MMP10 PubMed Links

Gene Details

Gene
MMP10
Gene Name
matrix metallopeptidase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002425.2 1540 UTR 3 NP_002416.1

View Full Product Details