Product Details

SNP ID
rs1076558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68039236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGCTCCGTGAATGCAGCTCAG[A/C]CCGCCACCCCGCAGGGTGTCAGGCC
Phenotype
MIM: 600466 MIM: 602141 MIM: 604592
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALDH3B1 PubMed Links

Gene Details

Gene
ALDH3B1
Gene Name
aldehyde dehydrogenase 3 family member B1
There are no transcripts associated with this gene.

Gene
MIR4691
Gene Name
microRNA 4691
There are no transcripts associated with this gene.

Gene
MIR6753
Gene Name
microRNA 6753
There are no transcripts associated with this gene.

Gene
MIR7113
Gene Name
microRNA 7113
There are no transcripts associated with this gene.

Gene
NDUFS8
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
There are no transcripts associated with this gene.

Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006019.3 Intron NP_006010.2
NM_006053.3 Intron NP_006044.1
XM_005273709.3 Intron XP_005273766.1
XM_011544726.2 Intron XP_011543028.1
XM_017017089.1 Intron XP_016872578.1
XM_017017090.1 Intron XP_016872579.1

View Full Product Details