Product Details

SNP ID
rs267731
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:150983106 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCCCCAGGGCAACCAGTAGCACC[A/G]TGTCTGTGACTGGCGGGAAGATGGC
Phenotype
MIM: 603319 MIM: 606920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA9 PubMed Links

Gene Details

Gene
ANXA9
Gene Name
annexin A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003568.2 190 Missense Mutation ATG,GTG M1V NP_003559.2
XM_011510058.2 190 Missense Mutation ATG,GTG M1V XP_011508360.1
Gene
CERS2
Gene Name
ceramide synthase 2
There are no transcripts associated with this gene.

View Full Product Details