Product Details

SNP ID
rs339723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42659138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCCAGGTGCCGGGCCAAGCCC[A/G]GCCACCGTGGTTCTGGGCCGGGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928323 PubMed Links

Gene Details

Gene
LOC101928323
Gene Name
uncharacterized LOC101928323
There are no transcripts associated with this gene.

Gene
ZBTB47
Gene Name
zinc finger and BTB domain containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145166.3 1064 Silent Mutation CCA,CCG P261P NP_660149.2

View Full Product Details