Product Details

SNP ID
rs1281378
Assay Type
Validated
NCBI dbSNP Submissions
84
Location
Chr.1:179882939 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CCTCCGCTACAGCCGTCTCCTGTTA[C/T]GACCAGGAGAGGGCTGCGGGACTCT
Phenotype
MIM: 614512 MIM: 614513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOR1AIP1 PubMed Links

Gene Details

Gene
TOR1AIP1
Gene Name
torsin 1A interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267578.1 898 Missense Mutation ACG,ATG T146M NP_001254507.1
NM_015602.3 898 Missense Mutation ACG,ATG T146M NP_056417.2
XM_011509403.1 898 Missense Mutation ACG,ATG T146M XP_011507705.1
XM_011509404.1 898 Missense Mutation ACG,ATG T146M XP_011507706.1
Gene
TOR1AIP2
Gene Name
torsin 1A interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details