Product Details
- SNP ID
-
rs34901329
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:48444908 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGATGCCAGCAGGAAAGCCAAAC[A/T]TCCATTATTTCTCTACAGTTTACTT
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP1R21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145713844] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP1R21
- Gene Name
- protein phosphatase 1 regulatory subunit 21
View Full Product Details