Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-1
Entrez Gene ID
181592
Gene Name
TBC (Tre-2/Bub2/Cdc16) domain family
Gene Aliases
CELE_F20D1.2
Chromosome Location
Chr.X: 14970751 - 14974780 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078084.7 NP_510485.2 9 - 10 1248 131

View Full Product Details