Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-27
Entrez Gene ID
174121
Gene Name
protein-coding
Gene Aliases
CELE_C04A2.3
Chromosome Location
Chr.II: 6835008 - 6847908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313565.3 NP_001300494.1 7 - 7 877 142
NM_001313568.3 NP_001300497.1 6 - 7 775 142
NM_001313569.3 NP_001300498.1 6 - 7 775 142
NM_001381462.1 NP_001368509.1 4 - 5 454 142
NM_001381464.1 NP_001368510.1 1 - 2 163 142
NM_001383871.1 7 - 8 921 142
NM_001383872.1 6 - 7 898 142
NM_001383873.1 7 - 8 943 142
NM_171011.4 NP_741012.1 7 - 8 937 142

View Full Product Details