Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
soc-2
Entrez Gene ID
177286
Gene Name
protein-coding
Gene Aliases
CELE_AC7.2
Chromosome Location
Chr.IV: 5127006 - 5152491 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026088.4 NP_001021259.1 1 - 2 268 72
GenBank mRNA AF054827.1 - 1 - 2 275 72
AF068919.1 - 1 - 2 181 72

View Full Product Details