Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
soc-2
Entrez Gene ID
177286
Gene Name
protein-coding
Gene Aliases
CELE_AC7.2
Chromosome Location
Chr.IV: 5127006 - 5152491 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026088.4 NP_001021259.1 7 - 8 1547 81
NM_001306829.3 NP_001293758.1 8 - 9 1700 81
NM_001306830.3 NP_001293759.1 8 - 9 1491 81
NM_171332.5 NP_741391.2 8 - 9 1465 81
GenBank mRNA AF054827.1 - 7 - 8 1554 81
AF068919.1 - 7 - 8 1460 81

View Full Product Details