Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckr-1
Entrez Gene ID
188774
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T23B3.4
Chromosome Location
Chr.I: 6687349 - 6691671 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059517.4 NP_491918.3 5 - 6 669 76

View Full Product Details