Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rig-5
Entrez Gene ID
172791
Gene Name
protein-coding
Gene Aliases
CELE_C36F7.4
Chromosome Location
Chr.I: 9565389 - 9574377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128955.4 NP_001122427.1 2 - 3 345 96
NM_001264200.4 NP_001251129.1 2 - 3 345 96
NM_001264202.1 NP_001251131.1 2 - 3 345 96
NM_001264203.2 NP_001251132.1 2 - 3 192 96
NM_001381092.1 NP_001366862.1 2 - 3 155 96
NM_060134.4 NP_492535.1 2 - 3 345 96

View Full Product Details