Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sep-1
Entrez Gene ID
171912
Gene Name
Separin homolog sep-1
Gene Aliases
CELE_Y47G6A.12
Chromosome Location
Chr.I: 3433155 - 3438639 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058759.6 NP_491160.1 3 - 4 960 63
GenBank mRNA AF399825.1 - 2 - 3 937 63

View Full Product Details