Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-57
Entrez Gene ID
172078
Gene Name
protein-coding
Gene Aliases
CELE_T04D1.3
Chromosome Location
Chr.I: 4675133 - 4679448 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047215.5 NP_001040680.1 4 - 5 815 69

View Full Product Details