Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
him-1
Entrez Gene ID
172116
Gene Name
protein-coding
Gene Aliases
CELE_F28B3.7
Chromosome Location
Chr.I: 4951570 - 4957667 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047193.5 NP_001040658.2 10 - 11 3579 74
NM_001047194.3 NP_001040659.1 1 - 2 206 74

View Full Product Details