Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-40
Entrez Gene ID
172233
Gene Name
Uncharacterized protein
Gene Aliases
CELE_T19B4.7
Chromosome Location
Chr.I: 5679837 - 5691503 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059263.10 NP_491664.1 15 - 16 3725 103
GenBank mRNA U70618.1 - 15 - 16 3667 103

View Full Product Details