Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-66
Entrez Gene ID
177618
Gene Name
protein-coding
Gene Aliases
CELE_T09A12.4
Chromosome Location
Chr.IV: 8226331 - 8236602 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307205.2 NP_001294134.1 1 - 2 92 73
NM_001383138.1 1 - 2 77 73
NM_171396.7 NP_741462.1 1 - 2 72 73

View Full Product Details