Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-66
Entrez Gene ID
177618
Gene Name
protein-coding
Gene Aliases
CELE_T09A12.4
Chromosome Location
Chr.IV: 8226331 - 8236602 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028198.5 NP_001023369.1 6 - 7 729 118
NM_001307204.3 NP_001294133.1 3 - 4 309 118
NM_001307205.2 NP_001294134.1 7 - 8 970 118
NM_001307206.3 NP_001294135.1 6 - 7 723 118
NM_001307207.3 NP_001294136.1 4 - 5 489 118
NM_001380356.1 NP_001368431.1 3 - 4 303 118
NM_001383138.1 7 - 8 1057 118
NM_001383139.1 3 - 4 559 118
NM_171396.7 NP_741462.1 7 - 8 956 118
NM_171397.8 NP_741463.1 4 - 5 483 118

View Full Product Details