Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dmd-7
Entrez Gene ID
179012
Gene Name
DM domain-containing protein
Gene Aliases
CELE_K08B12.2
Chromosome Location
Chr.V: 6249172 - 6252458 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171469.5 NP_741551.1 2 - 3 263 74

View Full Product Details