Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tnt-3
Entrez Gene ID
181052
Gene Name
protein-coding
Gene Aliases
CELE_C14F5.3
Chromosome Location
Chr.X: 7937633 - 7945099 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373379.1 NP_001360658.1 4 - 5 306 74
NM_001373380.2 NP_001359846.1 4 - 5 258 74
NM_001373381.2 NP_001359848.1 3 - 4 2933 74
NM_001373382.2 NP_001360657.1 3 - 4 2932 74
NM_001373383.1 NP_001359847.1 3 - 4 2933 74
NM_001383610.1 3 - 4 177 74

View Full Product Details