Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uba-2
Entrez Gene ID
24104970
Gene Name
SUMO-activating enzyme subunit uba-2
Gene Aliases
CELE_W02A11.4
Chromosome Location
Chr.I: 12741588 - 12749564 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306225.3 NP_001293154.1 5 - 6 593 71

View Full Product Details