Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lips-12
Entrez Gene ID
188468
Gene Name
LIPaSe related
Gene Aliases
CELE_T13B5.6
Chromosome Location
Chr.II: 1112184 - 1114233 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061514.3 NP_493915.1 3 - 4 660 81

View Full Product Details