Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rog-1
Entrez Gene ID
186242
Gene Name
protein-coding
Gene Aliases
CELE_F54D12.6
Chromosome Location
Chr.II: 1368102 - 1374989 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276728.3 NP_001263657.1 7 - 8 1416 77
NM_061557.3 NP_493958.2 7 - 8 1424 77

View Full Product Details