Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-28
Entrez Gene ID
185383
Gene Name
protein-coding
Gene Aliases
CELE_F36H5.3
Chromosome Location
Chr.II: 1763372 - 1766555 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047307.3 NP_001040772.1 5 - 6 948 62
NM_001047308.1 NP_001040773.1 5 - 6 948 62

View Full Product Details