Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-18
Entrez Gene ID
183351
Gene Name
MATH domain-containing protein
Gene Aliases
CELE_C40A11.1
Chromosome Location
Chr.II: 2154008 - 2155220 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061800.4 NP_494201.2 3 - 4 534 86

View Full Product Details