Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sri-29
- Entrez Gene ID
-
191904
- Gene Name
- Serpentine Receptor, class I
- Gene Aliases
-
CELE_B0454.4
- Chromosome Location
-
Chr.II: 3044323 - 3046115 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062030.1 |
NP_494431.1 |
3 - 4 |
757 |
67 |
- Gene Symbol
- sri-30
- Entrez Gene ID
-
191905
- Gene Name
- Serpentine Receptor, class I
- Gene Aliases
-
CELE_B0454.10
- Chromosome Location
-
Chr.II: 3044537 - 3048154 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062031.5 |
NP_494432.1 |
2 - 3 |
595 |
67 |
View Full Product Details