Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mdt-4
Entrez Gene ID
173852
Gene Name
Mediator of RNA polymerase II transcription subunit 4
Gene Aliases
CELE_ZK546.13
Chromosome Location
Chr.II: 4942107 - 4943949 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062504.5 NP_494905.2 4 - 5 833 83

View Full Product Details