Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-6
Entrez Gene ID
191777
Gene Name
Serpentine receptor class alpha-6
Gene Aliases
CELE_AH6.10
Chromosome Location
Chr.II: 9548405 - 9550818 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063651.2 NP_496052.1 2 - 3 1957 151

View Full Product Details