Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phg-1
Entrez Gene ID
185024
Gene Name
Growth arrest-specific protein 1 homolog
Gene Aliases
CELE_F27E5.4
Chromosome Location
Chr.II: 10137658 - 10139216 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063785.5 NP_496186.2 2 - 3 406 113

View Full Product Details