Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rol-1
Entrez Gene ID
174857
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_Y57A10A.11
Chromosome Location
Chr.II: 12172571 - 12176069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064188.5 NP_496589.1 3 - 4 359 113

View Full Product Details