Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sop-2
Entrez Gene ID
174874
Gene Name
protein-coding
Gene Aliases
CELE_C50E10.4
Chromosome Location
Chr.II: 12339944 - 12344799 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267411.2 NP_001254340.1 1 - 2 93 67
NM_001267412.2 NP_001254341.1 1 - 2 93 67

View Full Product Details