Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
moe-3
Entrez Gene ID
174960
Gene Name
CCCH-type zinc finger protein moe-3
Gene Aliases
CELE_F32A11.6
Chromosome Location
Chr.II: 13160411 - 13165733 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064394.3 NP_496795.1 2 - 3 316 64

View Full Product Details