Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-22
Entrez Gene ID
176231
Gene Name
protein-coding
Gene Aliases
CELE_PAR2.4
Chromosome Location
Chr.III: 8763534 - 8767341 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372637.3 NP_001360092.1 2 - 3 290 72
NM_066533.6 NP_498934.1 2 - 3 284 72
GenBank mRNA AB110823.1 - 1 - 2 347 72

View Full Product Details