Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- F54F2.14
- Entrez Gene ID
-
27219622
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F54F2.14
- Chromosome Location
-
Chr.III: 8791545 - 8806286 on Build WBcel235
- Gene Symbol
- zfp-1
- Entrez Gene ID
-
176237
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F54F2.2
- Chromosome Location
-
Chr.III: 8791545 - 8806289 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001382947.1 |
|
10 - 11 |
1949 |
64 |
NR_136383.1 |
|
9 - 10 |
1094 |
64 |
View Full Product Details