Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-7
Entrez Gene ID
176370
Gene Name
Putative sodium/calcium exchanger 7
Gene Aliases
CELE_C07A9.11
Chromosome Location
Chr.III: 9715881 - 9720291 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066746.7 NP_499147.3 11 - 12 1905 94

View Full Product Details