Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rle-1
Entrez Gene ID
176525
Gene Name
protein-coding
Gene Aliases
CELE_M142.6
Chromosome Location
Chr.III: 10936244 - 10947330 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379916.1 NP_001366782.1 3 - 4 537 116
NM_001379917.1 NP_001367296.1 3 - 4 539 116

View Full Product Details