Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
teg-1
Entrez Gene ID
176561
Gene Name
Tumorous Enhancer of Glp-1(gf)
Gene Aliases
CELE_Y47D3A.27
Chromosome Location
Chr.III: 11312213 - 11318231 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067054.7 NP_499455.2 4 - 5 874 105

View Full Product Details