Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pod-1
Entrez Gene ID
176784
Gene Name
protein-coding
Gene Aliases
CELE_Y76A2B.1
Chromosome Location
Chr.III: 13514836 - 13524122 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276804.3 NP_001263733.1 3 - 4 737 56
NM_001276805.4 NP_001263734.1 4 - 5 870 56
NM_001276806.2 NP_001263735.1 4 - 5 858 56
NM_067396.6 NP_499797.1 3 - 4 726 56
GenBank mRNA AF129126.2 - 3 - 4 748 56

View Full Product Details