Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pat-6
Entrez Gene ID
176848
Gene Name
Paralyzed arrest at two-fold protein 6
Gene Aliases
CELE_T21D12.4
Chromosome Location
Chr.IV: 261886 - 264232 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067490.6 NP_499891.1 2 - 3 248 78

View Full Product Details