Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnc-4
Entrez Gene ID
177600
Gene Name
DyNactin Complex component
Gene Aliases
CELE_C26B2.1
Chromosome Location
Chr.IV: 8018130 - 8019808 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068943.5 NP_501344.1 4 - 5 805 88

View Full Product Details