Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acdh-13
Entrez Gene ID
177654
Gene Name
Acyl CoA DeHydrogenase
Gene Aliases
CELE_C28C12.9
Chromosome Location
Chr.IV: 8496484 - 8498823 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069051.6 NP_501452.1 2 - 3 186 109

View Full Product Details