Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gtl-2
Entrez Gene ID
178036
Gene Name
Gon-Two Like (TRP subfamily)
Gene Aliases
CELE_F54D1.5
Chromosome Location
Chr.IV: 11273451 - 11281992 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069717.7 NP_502118.4 22 - 23 3890 69

View Full Product Details