Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asm-3
Entrez Gene ID
176879
Gene Name
protein-coding
Gene Aliases
CELE_W03G1.7
Chromosome Location
Chr.IV: 520214 - 523809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047531.2 NP_001040996.1 11 - 12 1416 109
NM_001047532.6 NP_001040997.1 10 - 11 1214 109

View Full Product Details