Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-70
Entrez Gene ID
185128
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_F30B5.6
Chromosome Location
Chr.IV: 4226858 - 4228017 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068115.1 NP_500516.1 1 - 2 181 72

View Full Product Details