Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-22
Entrez Gene ID
177369
Gene Name
protein-coding
Gene Aliases
CELE_C46G7.4
Chromosome Location
Chr.IV: 6019672 - 6033639 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306867.3 NP_001293796.1 4 - 5 1071 82
NM_001306868.2 NP_001293797.1 4 - 5 1071 82
NM_001306869.4 NP_001293798.1 4 - 5 1071 82
NM_001306870.4 NP_001293799.1 4 - 5 1071 82
NM_001306875.3 NP_001293804.1 5 - 6 1326 82
NM_001306876.3 NP_001293805.1 5 - 6 1326 82
NM_001306877.4 NP_001293806.1 5 - 6 1326 82
NM_068506.3 NP_500907.2 5 - 6 1326 82

View Full Product Details