Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrck-1
Entrez Gene ID
179013
Gene Name
protein-coding
Gene Aliases
CELE_K08B12.5
Chromosome Location
Chr.V: 6257390 - 6267287 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313212.2 NP_001300141.1 1 - 2 200 92
NM_072198.7 NP_504599.2 1 - 2 110 92

View Full Product Details