Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-70
Entrez Gene ID
179077
Gene Name
protein-coding
Gene Aliases
CELE_K11C4.3
Chromosome Location
Chr.V: 6881338 - 6891839 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028881.4 NP_001024052.1 10 - 11 6768 95
NM_001028882.5 NP_001024053.2 12 - 13 6903 95
NM_001269144.3 NP_001256073.1 9 - 10 6984 95
NM_001307743.4 NP_001294672.1 10 - 11 6800 95
GenBank mRNA AF166169.1 - 11 - 12 6851 95
AF166170.1 - 9 - 10 6716 95
AF261891.1 - 10 - 11 6784 95

View Full Product Details