Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-70
Entrez Gene ID
179077
Gene Name
protein-coding
Gene Aliases
CELE_K11C4.3
Chromosome Location
Chr.V: 6881338 - 6891839 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028881.4 NP_001024052.1 11 - 12 6862 89
NM_001028882.5 NP_001024053.2 13 - 14 6997 89
NM_001269144.3 NP_001256073.1 10 - 11 7078 89
NM_001307743.4 NP_001294672.1 11 - 12 6894 89
GenBank mRNA AF166169.1 - 12 - 13 6945 89
AF166170.1 - 10 - 11 6810 89
AF261891.1 - 11 - 12 6878 89

View Full Product Details